Skip to Content

BDPH9BCC4D4D

Bide Pharmatech

3-((6-Bromopyridin-2-yl)amino)propanoic acid

Synonym(s):

3-((6-bromopyridin-2-yl)amino)propanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H9O2N2Br1
CAS Number:
Molecular Weight:
245.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H9BrN2O2/c9-6-2-1-3-7(11-6)10-5-4-8(12)13/h1-3H,4-5H2,(H,10,11)(H,12,13)

Inchi Key

CECVZFZBEGRZND-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library