Skip to Content
Merck

BDPH9BCC5402

Bide Pharmatech

6-Bromo-1-(4-methylbenzenesulfonyl)-1H-indazole-4-carbonitrile

Synonym(s):

6-bromo-1-tosyl-1H-indazole-4-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C15H10O2N3Br1S1
CAS Number:
Molecular Weight:
376.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C15H10BrN3O2S/c1-10-2-4-13(5-3-10)22(20,21)19-15-7-12(16)6-11(8-17)14(15)9-18-19/h2-7,9H,1H3

Inchi Key

CCJHCPAOQAJKCH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library