Skip to Content
Merck

BDPH9BCC67E9

Bide Pharmatech

4-(Thiophen-3-yl)butanoic acid

Synonym(s):

4-(3-thienyl)butanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H10O2S1
CAS Number:
Molecular Weight:
170.23
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H10O2S/c9-8(10)3-1-2-7-4-5-11-6-7/h4-6H,1-3H2,(H,9,10)

Inchi Key

QICKSFILIHOSPO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Yellow liquid



Still not finding the right product?

Explore all of our products under 4-(Thiophen-3-yl)butanoic acid



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library