Skip to Content
Merck

BDPH9BCC6B61

Bide Pharmatech

1-Bromo-4-(hexyloxy)benzene

Synonym(s):

1-bromo-4-(hexyloxy)benzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H17O1Br1
CAS Number:
Molecular Weight:
257.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H17BrO/c1-2-3-4-5-10-14-12-8-6-11(13)7-9-12/h6-9H,2-5,10H2,1H3

Inchi Key

GKLMJONYGGTHHM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: liquid / Colour: Colorless - Pale-yellow



Still not finding the right product?

Explore all of our products under 1-Bromo-4-(hexyloxy)benzene


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library