Skip to Content
Merck

BDPH9BCC7AB1

Bide Pharmatech

5-Bromo-1-methyl-1H-pyrrole-2-carbaldehyde

Synonym(s):

5-bromo-1-methyl-1H-pyrrole-2-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N1Br1
CAS Number:
Molecular Weight:
188.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H6BrNO/c1-8-5(4-9)2-3-6(8)7/h2-4H,1H3

Inchi Key

LBKFOHZUSINQRU-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library