Skip to Content
Merck

BDPH9BCC7B92

Bide Pharmatech

4-Hydroxyphenyl methanesulfonate

Synonym(s):

4-hydroxyphenyl methanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H8O4S1
CAS Number:
Molecular Weight:
188.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H8O4S/c1-12(9,10)11-7-4-2-6(8)3-5-7/h2-5,8H,1H3

Inchi Key

SKJCKYVIQGBWTN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Hydroxyphenyl methanesulfonate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library