Skip to Content
Merck

BDPH9BCC9374

Bide Pharmatech

2-Bromo-1-fluorodibenzo[b,d]furan

Synonym(s):

12-bromo-13-fluoro-8-oxatricyclo[7.4.0.0²,⁷]trideca-1(9),2,4,6,10,12-hexaene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H6O1Br1F1
CAS Number:
Molecular Weight:
265.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H6BrFO/c13-8-5-6-10-11(12(8)14)7-3-1-2-4-9(7)15-10/h1-6H

Inchi Key

XHAMAQQPOFHFFR-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Bromo-1-fluorodibenzo[b,d]furan


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library