Skip to Content
Merck

BDPH9BCC95EB

Bide Pharmatech

2,2'-(1-Benzylpiperidine-4,4-diyl)diethanol

Synonym(s):

2,2'-(1-benzylpiperidine-4,4-diyl)diethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H25O2N1
CAS Number:
Molecular Weight:
263.38
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C16H25NO2/c18-12-8-16(9-13-19)6-10-17(11-7-16)14-15-4-2-1-3-5-15/h1-5,18-19H,6-14H2

Inchi Key

YKUVZEGONPYNPE-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library