Skip to Content
Merck

BDPH9BCC9A33

Bide Pharmatech

7-Chloro-3-methyl-1H-indole-2-carbaldehyde

Synonym(s):

7-chloro-3-methyl-1H-indole-2-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H8O1N1Cl1
Molecular Weight:
193.63
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H8ClNO/c1-6-7-3-2-4-8(11)10(7)12-9(6)5-13/h2-5,12H,1H3

Inchi Key

FLYFADJEUDJOJU-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library