Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H9N3O2/c1-2-14-9(13)7-6-8-10-4-3-5-12(8)11-7/h3-6H,2H2,1H3
Inchi Key
HDHWFJFNCDMGJK-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
White to Yellow Solid
Still not finding the right product?
Explore all of our products under Ethyl pyrazolo[1,5-a]pyrimidine-2-carboxylate
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
