Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C6H5BrFNO/c1-10-5-3-2-4(7)6(8)9-5/h2-3H,1H3
Inchi Key
MQTONQLSSAOFRP-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Solid or Semi-solid or liquid or lump
Still not finding the right product?
Explore all of our products under 3-Bromo-2-fluoro-6-methoxypyridine
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
