Skip to Content
Merck

BDPH9BCCCCBD

Bide Pharmatech

1-Formylcyclopropane-1-carbonitrile

Synonym(s):

1-formylcyclopropane-1-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H5O1N1
CAS Number:
Molecular Weight:
95.1
MDL number:
Product BDPH9BCCCCBD is not currently sold in your country.


Inchi Code

1S/C5H5NO/c6-3-5(4-7)1-2-5/h4H,1-2H2

Inchi Key

ZVJZAKIRDRMLDW-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library