Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C7H5BrN2S/c1-4-2-3-5(8)7-6(4)9-11-10-7/h2-3H,1H3
Inchi Key
MZFRKYXVQYNOFA-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, 2-8C
Country of Origin
CN
Physical Form
Light yellow to brown powder or crystals
Still not finding the right product?
Explore all of our products under 4-Bromo-7-methylbenzo[c][1,2,5]thiadiazole
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
