Skip to Content
Merck

BDPH9BCCE2FB

Bide Pharmatech

3-Bromo-4-chloro-2-fluoroaniline

Synonym(s):

3-bromo-4-chloro-2-fluoroaniline

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H4N1Cl1Br1F1
CAS Number:
Molecular Weight:
224.46
MDL number:
Product BDPH9BCCE2FB is not currently sold in your country.


Inchi Code

1S/C6H4BrClFN/c7-5-3(8)1-2-4(10)6(5)9/h1-2H,10H2

Inchi Key

NNEAPESSFSAYBJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

White to Yellow to Brown Solid

Storage temp.

Keep in dark place, sealed in dry, 2-8°C



Still not finding the right product?

Explore all of our products under 3-Bromo-4-chloro-2-fluoroaniline


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library