Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C9H10BrFO/c10-8-4-3-7(2-1-5-12)9(11)6-8/h3-4,6,12H,1-2,5H2
Inchi Key
FCONJJUHDSNWII-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
98%
Storage temp.
Sealed in dry, room temperature
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 3-(4-Bromo-2-fluorophenyl)propan-1-ol
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
