Skip to Content
Merck

BDPH9BCCE5EB

Bide Pharmatech

6-Amino-5-chloropicolinic acid

Synonym(s):

6-amino-5-chloro-2-pyridinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O2N2Cl1
CAS Number:
Molecular Weight:
172.57
MDL number:
Product BDPH9BCCE5EB is not currently sold in your country.


Inchi Code

1S/C6H5ClN2O2/c7-3-1-2-4(6(10)11)9-5(3)8/h1-2H,(H2,8,9)(H,10,11)

Inchi Key

KBCHPUZXYDMCMN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?

Explore all of our products under 6-Amino-5-chloropicolinic acid


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library