Skip to Content

BDPH9BCCEF7A

Bide Pharmatech

Ethyl 3-(butylamino)propanoate

Synonym(s):

ethyl 3-(butylamino)propanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H19O2N1
CAS Number:
Molecular Weight:
173.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H19NO2/c1-3-5-7-10-8-6-9(11)12-4-2/h10H,3-8H2,1-2H3

Inchi Key

HTFPNDKVPVWBDL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?

Explore all of our products under Ethyl 3-(butylamino)propanoate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library