Skip to Content
Merck

BDPH9BCCF9C7

Bide Pharmatech

7-Methyl-2,3-dihydrobenzo[b][1,4]dioxine-6-carbaldehyde

Synonym(s):

7-methyl-2,3-dihydro-1,4-benzodioxine-6-carbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H10O3
CAS Number:
Molecular Weight:
178.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H10O3/c1-7-4-9-10(5-8(7)6-11)13-3-2-12-9/h4-6H,2-3H2,1H3

Inchi Key

HQDFDCPSGBNTRJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library