Skip to Content
Merck

BDPH9BCCFB1A

Bide Pharmatech

3-Bromo-6-chloro-2-iodopyridine

Synonym(s):

3-bromo-6-chloro-2-iodopyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H2N1Cl1Br1I1
CAS Number:
Molecular Weight:
318.34
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H2BrClIN/c6-3-1-2-4(7)9-5(3)8/h1-2H

Inchi Key

PNKGIZKPKHAJOI-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 3-Bromo-6-chloro-2-iodopyridine


Signal Word

Danger

Pictograms

CorrosionSkull and crossbones


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library