Skip to Content

BDPH9BCCFDD6

Bide Pharmatech

Thiazole-5-carbonitrile

Synonym(s):

1,3-thiazole-5-carbonitrile

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H2N2S1
CAS Number:
Molecular Weight:
110.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H2N2S/c5-1-4-2-6-3-7-4/h2-3H

Inchi Key

XWXPRCDFFCVMNT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Thiazole-5-carbonitrile


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library