Inchi Code
1S/C8H8FNO2/c1-12-8(11)6-3-2-5(9)4-7(6)10/h2-4H,10H2,1H3
Inchi Key
UBFRSTYHLYPSND-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Solid
Still not finding the right product?
Explore all of our products under Methyl 2-amino-4-fluorobenzoate
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
