Skip to Content
Merck

BDPH9BCD08AE

Bide Pharmatech

4-Bromo-2,6-dimethylbenzaldehyde

Synonym(s):

4-bromo-2,6-dimethylbenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H9O1Br1
CAS Number:
Molecular Weight:
213.07
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H9BrO/c1-6-3-8(10)4-7(2)9(6)5-11/h3-5H,1-2H3

Inchi Key

JFRPWJDOGGLZFS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 4-Bromo-2,6-dimethylbenzaldehyde


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library