Skip to Content
Merck

BDPH9BCD1398

Bide Pharmatech

5-Methyl-3-(2'-chlorophenyl)-4-isoxazolecarboxylic acid

Synonym(s):

3-(2-chlorophenyl)-5-methyl-4-isoxazolecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H8O3N1Cl1
Molecular Weight:
237.64
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H8ClNO3/c1-6-9(11(14)15)10(13-16-6)7-4-2-3-5-8(7)12/h2-5H,1H3,(H,14,15)

Inchi Key

UVEPOHNXGXVOJE-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library