Skip to Content
Merck

BDPH9BCD13A2

Bide Pharmatech

5-Bromo-3-hydroxyquinolin-2(1H)-one

Synonym(s):

5-bromo-3-hydroxy-2(1H)-quinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H6O2N1Br1
CAS Number:
Molecular Weight:
240.06
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H6BrNO2/c10-6-2-1-3-7-5(6)4-8(12)9(13)11-7/h1-4,12H,(H,11,13)

Inchi Key

RXMNJPQCLAHRIZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library