Skip to Content
Merck

BDPH9BCD16EC

Bide Pharmatech

2-Cyano-N-(prop-2-yn-1-yl)acetamide

Synonym(s):

2-cyano-N-(2-propynyl)acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H6O1N2
CAS Number:
Molecular Weight:
122.13
MDL number:
Product BDPH9BCD16EC is not currently sold in your country.


Inchi Code

1S/C6H6N2O/c1-2-5-8-6(9)3-4-7/h1H,3,5H2,(H,8,9)

Inchi Key

UWMHFSKISKQAEA-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library