Skip to Content
Merck

BDPH9BCD17DA

Bide Pharmatech

3-Chloro-5-(difluoromethoxy)benzoic acid

Synonym(s):

3-chloro-5-(difluoromethoxy)benzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O3Cl1F2
CAS Number:
Molecular Weight:
222.58
MDL number:
Product BDPH9BCD17DA is not currently sold in your country.


Inchi Code

1S/C8H5ClF2O3/c9-5-1-4(7(12)13)2-6(3-5)14-8(10)11/h1-3,8H,(H,12,13)

Inchi Key

WOQULMFOHBBORM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Country of Origin

CN

Physical Form

Solid or liquid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library