Skip to Content

BDPH9BCD2131

Bide Pharmatech

4,7-Difluoro-1H-indole

Synonym(s):

4,7-difluoro-1H-indole

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5N1F2
CAS Number:
Molecular Weight:
153.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5F2N/c9-6-1-2-7(10)8-5(6)3-4-11-8/h1-4,11H

Inchi Key

WXDUAQYBARJXOQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or lump or liquid



Still not finding the right product?

Explore all of our products under 4,7-Difluoro-1H-indole


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library