Skip to Content

BDPH9BCD2A64

Bide Pharmatech

3-Cyclobutyl-3,9-diazaspiro[5.5]undecane

Synonym(s):

3-cyclobutyl-3,9-diazaspiro[5.5]undecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H24N2
CAS Number:
Molecular Weight:
208.35
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H24N2/c1-2-12(3-1)15-10-6-13(7-11-15)4-8-14-9-5-13/h12,14H,1-11H2

Inchi Key

LROBDXNVZFCHGC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid or semi-solid or liquid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library