Skip to Content
Merck

BDPH9BCD2F48

Bide Pharmatech

tert-Butyl 3-hydroxyazepane-1-carboxylate

Synonym(s):

tert-butyl 3-hydroxy-1-azepanecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H21O3N1
CAS Number:
Molecular Weight:
215.29
MDL number:
Product BDPH9BCD2F48 is not currently sold in your country.


Inchi Code

1S/C11H21NO3/c1-11(2,3)15-10(14)12-7-5-4-6-9(13)8-12/h9,13H,4-8H2,1-3H3

Inchi Key

YLXNIRRSRXUTCK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Colorless to Yellow Sticky Oil to Semi-Solid to Solid



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library