Skip to Content
Merck

BDPH9BCD372A

Bide Pharmatech

4-(2-Fluoro-5-methoxybenzyl)piperidine hydrochloride

Synonym(s):

4-(2-fluoro-5-methoxybenzyl)piperidine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C13H19O1N1Cl1F1
CAS Number:
Molecular Weight:
259.75
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C13H18FNO.ClH/c1-16-12-2-3-13(14)11(9-12)8-10-4-6-15-7-5-10;/h2-3,9-10,15H,4-8H2,1H3;1H

Inchi Key

LMPNEPFUDVRURO-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library