Inchi Code
1S/C7H6ClN3/c8-7-5-2-1-4(9)3-6(5)10-11-7/h1-3H,9H2,(H,10,11)
Inchi Key
DRRARKIFTNKQDW-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Light-red to yellow-brown solid
Still not finding the right product?
Explore all of our products under 6-Amino-3-chloro-1H-indazole
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
