Skip to Content
Merck

BDPH9BCD4007

Bide Pharmatech

Ethyl 2,4-dichloro-3-oxobutanoate

Synonym(s):

ethyl 2,4-dichloro-3-oxobutanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O3Cl2
CAS Number:
Molecular Weight:
199.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C6H8Cl2O3/c1-2-11-6(10)5(8)4(9)3-7/h5H,2-3H2,1H3

Inchi Key

GPDFVBIDCBPHSW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under Ethyl 2,4-dichloro-3-oxobutanoate



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library