Skip to Content

BDPH9BCD4480

Bide Pharmatech

N-(1-Phenylethyl)formamide

Synonym(s):

1-phenylethylformamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H11O1N1
Molecular Weight:
149.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H11NO/c1-8(10-7-11)9-5-3-2-4-6-9/h2-8H,1H3,(H,10,11)

Inchi Key

CDHCCWRMWKZBGE-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library