Skip to Content
Merck

Skip To

BDPH9BCD5008

Bide Pharmatech

N-Cyclopropyl-4-fluorobenzenesulfonamide

Synonym(s):

N-cyclopropyl-4-fluorobenzenesulfonamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C9H10O2N1S1F1
CAS Number:
Molecular Weight:
215.25
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10FNO2S/c10-7-1-5-9(6-2-7)14(12,13)11-8-3-4-8/h1-2,5-6,8,11H,3-4H2

Inchi Key

QXHJRYPFPZGNNV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library