Skip to Content

BDPH9BCD5C99

Bide Pharmatech

7-(Aminomethyl)indole

Synonym(s):

1H-indol-7-ylmethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10N2
CAS Number:
Molecular Weight:
146.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10N2/c10-6-8-3-1-2-7-4-5-11-9(7)8/h1-5,11H,6,10H2

Inchi Key

NMNYQFZHZZSSQC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 7-(Aminomethyl)indole




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library