Skip to Content
Merck

Skip To

BDPH9BCD65DA

Bide Pharmatech

3-Amino-5-(methylthio)pentanoic acid

Synonym(s):

3-amino-5-(methylsulfanyl)pentanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H13O2N1S1
CAS Number:
Molecular Weight:
163.24
MDL number:
Product BDPH9BCD65DA is not currently sold in your country.


Inchi Code

1S/C6H13NO2S/c1-10-3-2-5(7)4-6(8)9/h5H,2-4,7H2,1H3,(H,8,9)

Inchi Key

QWVNCDVONVDGDV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library