Skip to Content
Merck

BDPH9BCD6A4F

Bide Pharmatech

Methyl 4-amino-6-chloronicotinate

Synonym(s):

methyl 4-amino-6-chloronicotinate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O2N2Cl1
CAS Number:
Molecular Weight:
186.6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7ClN2O2/c1-12-7(11)4-3-10-6(8)2-5(4)9/h2-3H,1H3,(H2,9,10)

Inchi Key

CMIKPJBQKHVWJH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under Methyl 4-amino-6-chloronicotinate


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library