Skip to Content

BDPH9BCD6C44

Bide Pharmatech

4-(2-Aminoethyl)-4H-1,2,4-triazole

Synonym(s):

2-(4H-1,2,4-triazol-4-yl)ethanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C4H8N4
CAS Number:
Molecular Weight:
112.13
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H8N4/c5-1-2-8-3-6-7-4-8/h3-4H,1-2,5H2

Inchi Key

ATCZRYPJBPXFIN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 4-(2-Aminoethyl)-4H-1,2,4-triazole


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library