Skip to Content
Merck

BDPH9BCD7DBA

Bide Pharmatech

1-Fluoro-2,6-bis(methoxycarbonyl)pyridin-1-ium trifluoromethanesulfonate

Synonym(s):

1-fluoro-2,6-bis(methoxycarbonyl)pyridin-1-ium trifluoromethanesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O7N1S1F4
CAS Number:
Molecular Weight:
363.24
MDL number:
Product BDPH9BCD7DBA is not currently sold in your country.


Inchi Code

1S/C9H9FNO4.CHF3O3S/c1-14-8(12)6-4-3-5-7(11(6)10)9(13)15-2;2-1(3,4)8(5,6)7/h3-5H,1-2H3;(H,5,6,7)/q+1;/p-1

Inchi Key

LDTLFVAJBQGQPV-UHFFFAOYSA-M

Shipping Temperature

Cold-chain transportation

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library