Skip to Content
Merck

BDPH9BCD80C4

Bide Pharmatech

8-Methoxyquinoline-4-carboxylic acid

Synonym(s):

8-methoxy-4-quinolinecarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H9O3N1
CAS Number:
Molecular Weight:
203.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H9NO3/c1-15-9-4-2-3-7-8(11(13)14)5-6-12-10(7)9/h2-6H,1H3,(H,13,14)

Inchi Key

NEBCNIRZMNKUQQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Country of Origin

CN

Physical Form

White to Yellow Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library