Skip to Content

BDPH9BCD8F71

Bide Pharmatech

(3-(2-Ethoxy-2-oxoethyl)phenyl)boronic acid

Synonym(s):

3-(2-ethoxy-2-oxoethyl)phenylboronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H13O4B1
CAS Number:
Molecular Weight:
208.02
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H13BO4/c1-2-15-10(12)7-8-4-3-5-9(6-8)11(13)14/h3-6,13-14H,2,7H2,1H3

Inchi Key

GDWZKERCECMYJX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

White to Yellow Solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library