Skip to Content
Merck

BDPH9BCD9A67

Bide Pharmatech

(2-(Methoxycarbonyl)pyrimidin-5-yl)boronic acid

Synonym(s):

(2-(methoxycarbonyl)pyrimidin-5-yl)boronic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H7O4N2B1
CAS Number:
Molecular Weight:
181.94
MDL number:
Product BDPH9BCD9A67 is not currently sold in your country.


Inchi Code

1S/C6H7BN2O4/c1-13-6(10)5-8-2-4(3-9-5)7(11)12/h2-3,11-12H,1H3

Inchi Key

QJBIYMZDLMVGCM-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library