Skip to Content
Merck

BDPH9BCDA85F

Bide Pharmatech

2,3-Dibromo-4-fluorobenzaldehyde

Synonym(s):

2,3-dibromo-4-fluorobenzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H3O1Br2F1
CAS Number:
Molecular Weight:
281.91
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H3Br2FO/c8-6-4(3-11)1-2-5(10)7(6)9/h1-3H

Inchi Key

KSUPOCPXYXSEAP-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under 2,3-Dibromo-4-fluorobenzaldehyde



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library