Skip to Content
Merck

BDPH9BCDAFEE

Bide Pharmatech

8-Chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Synonym(s):

8-chloro-6-(trifluoromethyl)imidazo[1,2-a]pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H4N2Cl1F3
CAS Number:
Molecular Weight:
220.58
MDL number:
Product BDPH9BCDAFEE is not currently sold in your country.


Inchi Code

1S/C8H4ClF3N2/c9-6-3-5(8(10,11)12)4-14-2-1-13-7(6)14/h1-4H

Inchi Key

BWSHPRLSCABXRL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

96%

Country of Origin

CN

Physical Form

Solid

Storage temp.

Sealed in dry, 2-8°C



Still not finding the right product?


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library