Skip to Content
Merck

BDPH9BCDC0D2

Bide Pharmatech

2-Chloro-4-(4-(dibenzo[b,d]furan-4-yl)phenyl)-6-phenyl-1,3,5-triazine

Synonym(s):

2-chloro-4-(4-(dibenzo[b,d]furan-4-yl)phenyl)-6-phenyl-1,3,5-triazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C27H16O1N3Cl1
CAS Number:
Molecular Weight:
433.9
MDL number:
Product BDPH9BCDC0D2 is not currently sold in your country.


Inchi Code

1S/C27H16ClN3O/c28-27-30-25(18-7-2-1-3-8-18)29-26(31-27)19-15-13-17(14-16-19)20-10-6-11-22-21-9-4-5-12-23(21)32-24(20)22/h1-16H

Inchi Key

QODQTZSWXYTAMS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library