Skip to Content

BDPH9BCDC44B

Bide Pharmatech

Tris(4-bromo-2,3,5,6-tetramethylphenyl)borane

Synonym(s):

tris(4-bromo-2,3,5,6-tetramethylphenyl)borane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C30H36Br3B1
CAS Number:
Molecular Weight:
647.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C30H36BBr3/c1-13-19(7)28(32)20(8)14(2)25(13)31(26-15(3)21(9)29(33)22(10)16(26)4)27-17(5)23(11)30(34)24(12)18(27)6/h1-12H3

Inchi Key

CDDHNGCDCAKILA-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

95%

Storage temp.

Sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

Flame


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library