Skip to Content
Merck

BDPH9BCDC4FA

Bide Pharmatech

tert-Butyl 4-(1-aminopropan-2-yl)piperazine-1-carboxylate

Synonym(s):

tert-butyl 4-(2-amino-1-methylethyl)-1-piperazinecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H25O2N3
CAS Number:
Molecular Weight:
243.35
MDL number:
Product BDPH9BCDC4FA is not currently sold in your country.


Inchi Code

1S/C12H25N3O2/c1-10(9-13)14-5-7-15(8-6-14)11(16)17-12(2,3)4/h10H,5-9,13H2,1-4H3

Inchi Key

BYCHAILZTCZXAW-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library