Skip to Content

BDPH9BCDC50F

Bide Pharmatech

2-Bromo-6-(difluoromethoxy)benzaldehyde

Synonym(s):

2-bromo-6-(difluoromethoxy)benzaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H5O2Br1F2
CAS Number:
Molecular Weight:
251.03
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H5BrF2O2/c9-6-2-1-3-7(5(6)4-12)13-8(10)11/h1-4,8H

Inchi Key

YIDKFKJYJWBGKX-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

98%

Storage temp.

Inert atmosphere, store in freezer, under -20C

Country of Origin

CN

Physical Form

White to yellow to brown solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library