Skip to Content
Merck

Skip To

BDPH9BCDC66F

Bide Pharmatech

3-(Piperidin-1-yl)propan-1-amine

Synonym(s):

3-(1-piperidinyl)propylamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C8H18N2
CAS Number:
Molecular Weight:
142.24
MDL number:
Pricing and availability is not currently available. Contact Technical Service
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C8H18N2/c9-5-4-8-10-6-2-1-3-7-10/h1-9H2

Inchi Key

JMUCXULQKPWSTJ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: liquid



Still not finding the right product?

Explore all of our products under 3-(Piperidin-1-yl)propan-1-amine


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

CorrosionExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library