Skip to Content
Merck

BDPH9BCDCEC2

Bide Pharmatech

1,1,1,3,3,5,5,7,7,9,9,11,11,13,13,13-Hexadecamethylheptasiloxane

Synonym(s):

1,1,1,3,3,5,5,7,7,9,9,11,11,13,13,13-hexadecamethylheptasiloxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H48O6Si7
CAS Number:
Molecular Weight:
533.15
MDL number:
Product BDPH9BCDCEC2 is not currently sold in your country.


Inchi Code

1S/C16H48O6Si7/c1-23(2,3)17-25(7,8)19-27(11,12)21-29(15,16)22-28(13,14)20-26(9,10)18-24(4,5)6/h1-16H3

Inchi Key

NFVSFLUJRHRSJG-UHFFFAOYSA-N

Shipping Temperature

Cold-chain transportation

Purity

96%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Colorless to Yellow Liquid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library